1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-5-Oxo-Pro-OH

A483768-100MG A483768-250MG A483768-1G
77265-506EA 51.5 USD
77265-506 77265-508 77617-096
1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₅
MW: 229.22 g/mol
MDL Number: MFCD02308236
CAS Number: 160401-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR