3-Chloro-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A483120-10G A483120-1G A483120-250MG
77241-350EA 136.99 USD
77241-350 77241-352 77241-354
3-Chloro-N,N-dimethylbenzamide 98%
3-Chloro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
MDL Number: MFCD01214035
CAS Number: 24167-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR