2-Amino-3-(methoxycarbonyl)benzoic acid 98%

Supplier: Ambeed
Warning

A482860-100MG A482860-250MG A482860-5G A482860-1G
77279-504EA 20 USD
77279-504 77279-506 77748-998 77748-997
2-Amino-3-(methoxycarbonyl)benzoic acid 98%
2-Amino-3-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD06200897
CAS Number: 253120-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR