(R)-(+)-Etomoxir 98%

Supplier: Ambeed
Warning

A482603-10MG A482603-1MG A482603-25MG A482603-5MG A482603-50MG A482603-100MG A482603-250MG
77241-342EA 57.01 USD
77241-342 77241-344 77241-346 77241-348 77617-092 77746-991 77746-992
(R)-(+)-Etomoxir 98%
(R)-(+)-Etomoxir

Precaución: for Research Use Only

Formula: C₁₇H₂₃ClO₄
MDL Number: MFCD11227266
CAS Number: 124083-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR