Potassium 2-(quinolin-4-yl)acetate 95%

Supplier: Ambeed
Warning

A482436-100MG A482436-1G
77265-498EA 106.09 USD
77265-498 77265-500
Potassium 2-(quinolin-4-yl)acetate 95%
Potassium 2-(quinolin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₈KNO₂
MW: 225.28 g/mol
MDL Number: MFCD29905526
CAS Number: 2007920-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR