Benzyl 2-(4-hydroxyphenyl)acetate 95+%

Supplier: Ambeed
Warning

A482007-5G A482007-1G
77291-946EA 125.66 USD
77291-946 77748-996
Benzyl 2-(4-hydroxyphenyl)acetate 95+%
Benzyl 2-(4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00189402
CAS Number: 27727-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR