3-Hydroxy-3-methylbutanenitrile 97%

Supplier: Ambeed
Warning

A481357-1G A481357-25G A481357-5G
77279-492EA 17 USD
77279-492 77279-494 77279-496
3-Hydroxy-3-methylbutanenitrile 97%
3-Hydroxy-3-methylbutanenitrile

Precaución: for Research Use Only

Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD00043123
CAS Number: 13635-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR