2-Chloro-6-methoxynicotinic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-6-methoxypyridine-3-carboxylic acid
A481347-100MG
A481347-1G
A481347-250MG
A481347-5G
77265-490EA
13
USD
77265-490
77265-492
77265-494
77265-496
2-Chloro-6-methoxynicotinic acid 95%
2-Chloro-6-methoxynicotinic acid
Precaución: for Research Use Only
Formula:
C₇H₆NO₃Cl |
MDL Number:
MFCD08236845 CAS Number: 503000-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...