3-Methyl-1,5-pentanediol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methylpentane-1,5-diol

A481201-1000G A481201-100G A481201-500G
77279-486EA 69.01 USD
77279-486 77279-488 77279-490
3-Methyl-1,5-pentanediol 98%
3-Methyl-1,5-pentanediol

Precaución: for Research Use Only

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
MDL Number: MFCD00039547
CAS Number: 4457-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR