2,4-Dihydroxybenzhydrazide 98%

Supplier: Ambeed
Warning

A480915-250MG A480915-25G A480915-5G
77253-134EA 12 USD
77253-134 77253-136 77253-138
2,4-Dihydroxybenzhydrazide 98%
2,4-Dihydroxybenzhydrazide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD00017053
CAS Number: 13221-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR