2-Pentadecanone 95%

Supplier: Ambeed
Warning

Synonyms: Pentadecan-2-one

A480812-25G A480812-5G
77279-474EA 86.52 USD
77279-474 77279-476
2-Pentadecanone 95%
2-Pentadecanone

Precaución: for Research Use Only

Formula: C₁₅H₃₀O
MW: 226.4 g/mol
MDL Number: MFCD00053712
CAS Number: 2345-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR