Ethyl 3,3-difluoro-2-oxoindoline-5-carboxylate 97%

Supplier: Ambeed
Warning

A480757-100MG A480757-1G A480757-250MG
77241-336EA 98.88 USD
77241-336 77241-338 77241-340
Ethyl 3,3-difluoro-2-oxoindoline-5-carboxylate 97%
Ethyl 3,3-difluoro-2-oxoindoline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉F₂NO₃
MW: 241.19 g/mol
CAS Number: 1286793-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR