2-Ethylhexyl cyanoacetate 98%

Supplier: Ambeed
Warning

A479578-100G A479578-25G A479578-5G
77265-474EA 38 USD
77265-474 77265-476 77265-478
2-Ethylhexyl cyanoacetate 98%
2-Ethylhexyl cyanoacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₂
MDL Number: MFCD00019843
CAS Number: 13361-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR