7-Methoxyindole 97%

Supplier: Ambeed
Warning

A479405-1G A479405-25G A479405-5G
77279-466EA 10 USD
77279-466 77279-468 77279-470
7-Methoxyindole 97%
7-Methoxyindole

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00047203
CAS Number: 3189-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR