4-Ethoxy-3-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A478882-100G A478882-10G A478882-1G A478882-25G A478882-5G
77279-456EA 210 USD
77279-456 77279-458 77279-460 77279-462 77279-464
4-Ethoxy-3-fluorophenylboronic acid 98%
4-Ethoxy-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD04115667
CAS Number: 279263-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR