(S)-tert-Butyl (3-oxocyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

A478593-100MG A478593-250MG A478593-1G
77265-462EA 15 USD
77265-462 77265-464 77748-992
(S)-tert-Butyl (3-oxocyclohexyl)carbamate 97%
(S)-tert-Butyl (3-oxocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD30181862
CAS Number: 1803033-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR