2-Bromoethyl acrylate 98%

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A478367-5G A478367-25G
77265-460EA 69 USD
77265-460 77594-494
2-Bromoethyl acrylate 98%
2-Bromoethyl acrylate

Precaución: for Research Use Only

Formula: CH₂=CHCO₂CH₂CH₂Br
MDL Number: MFCD00013541
CAS Number: 4823-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR