N-(2-Chloro-5-(trifluoromethyl)phenyl)-2-(4-(furan-2-carbonyl)piperazin-1-yl)acetamide 99+%

Supplier: Ambeed
Warning

Synonyms: ML348

A477540-10MG A477540-1MG A477540-25MG A477540-50MG A477540-5MG
77265-444EA 23.69 USD
77265-444 77265-446 77265-448 77265-450 77265-452
N-(2-Chloro-5-(trifluoromethyl)phenyl)-2-(4-(furan-2-carbonyl)piperazin-1-yl)acetamide 99+%
N-(2-Chloro-5-(trifluoromethyl)phenyl)-2-(4-(furan-2-carbonyl)piperazin-1-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₈H₁₇ClF₃N₃O₃
MW: 415.79 g/mol
CAS Number: 899713-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR