(3-Bromopropyl)trimethoxysilane 98%

Supplier: Ambeed
Warning

Synonyms: Trimethoxy(3-bromopropyl)silane, 3-(Trimethoxysilyl)propylbromide

A477508-1G A477508-5G
77279-446EA 16.48 USD
77279-446 77279-448
(3-Bromopropyl)trimethoxysilane 98%
(3-Bromopropyl)trimethoxysilane

Precaución: for Research Use Only

Formula: BrH₂CCH₂CH₂Si(OCH₃)₃
MW: 243.17 g/mol
MDL Number: MFCD00053894
CAS Number: 51826-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR