4,4'-(ɑ-Methylbenzylidene)bisphenol 99%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(1-Phenylethylidene)diphenol, 1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 4,4-(1-Phenylethane-1,1-diyl)diphenol

A476765-100G A476765-25G A476765-500G A476765-5G
77291-924EA 51.5 USD
77291-924 77291-926 77291-928 77291-930
4,4'-(ɑ-Methylbenzylidene)bisphenol 99%
4,4'-(alpha-Methylbenzylidene)bisphenol

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₂
MW: 290.36 g/mol
MDL Number: MFCD00134687
CAS Number: 1571-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR