3-(Methylamino)propanoic acid 98%

Supplier: Ambeed
Danger

Synonyms: N-Methyl-β-alanine

A476262-1G A476262-5G
77279-422EA 23 USD
77279-422 77279-424
3-(Methylamino)propanoic acid 98%
3-(Methylamino)propanoic acid

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD06208295
CAS Number: 2679-14-3
UN: 2923
ADR: 8,6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR