2-Amino-4-methoxy-6-methyl-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A475808-10G A475808-25G A475808-500G A475808-100G
77279-416EA 5 USD
77279-416 77279-418 77279-420 77748-986
2-Amino-4-methoxy-6-methyl-1,3,5-triazine 97%
2-Amino-4-methoxy-6-methyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₅H₈N₄O
MW: 140.1400 g/mol
MDL Number: MFCD00052764
CAS Number: 1668-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR