2-Methyl-3-nitroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A475626-100MG A475626-1G A475626-250MG A475626-5G
77279-408EA 51.5 USD
77279-408 77279-410 77279-412 77279-414
2-Methyl-3-nitroimidazo[1,2-a]pyridine 97%
2-Methyl-3-nitroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MDL Number: MFCD09800612
CAS Number: 34165-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR