2,2,3,3-Tetrafluoro-1,4-butanediol 98%

Supplier: Ambeed
Warning

Synonyms: 2,2,3,3-Tetrafluorobutane-1,4-diol

A475440-1G A475440-25G A475440-5G
77253-084EA 29.87 USD
77253-084 77253-086 77253-088
2,2,3,3-Tetrafluoro-1,4-butanediol 98%
2,2,3,3-Tetrafluoro-1,4-butanediol

Precaución: for Research Use Only

Formula: C₄H₆F₄O₂
MW: 162.08 g/mol
MDL Number: MFCD00042375
CAS Number: 425-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR