4-(2-Morpholin-4-ylethoxy)phenylamine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Morpholinoethoxy)aniline, 4-[2-(4-Aminophenoxy)ethyl]morpholine, 4-[2-(4-Morpholinyl)ethoxy]aniline

A475383-100MG A475383-1G A475383-250MG A475383-5G
77291-902EA 10.3 USD
77291-902 77291-904 77291-906 77291-908
4-(2-Morpholin-4-ylethoxy)phenylamine 97%
4-(2-Morpholin-4-ylethoxy)phenylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD04970981
CAS Number: 52481-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR