(R)-tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A475212-100MG A475212-1G A475212-250MG A475212-5G
77265-418EA 98.88 USD
77265-418 77265-420 77265-422 77572-496
(R)-tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate 98%
(R)-tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆FNO₃
MW: 217.24 g/mol
MDL Number: MFCD28010938
CAS Number: 1266339-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR