Trimethoxy(4-methoxyphenyl)silane 93%

Supplier: Ambeed
Warning

Synonyms: 4-(Trimethoxysilyl)anisole

A475148-100MG A475148-1G A475148-250MG A475148-5G
77291-894EA 28 USD
77291-894 77291-896 77291-898 77291-900
Trimethoxy(4-methoxyphenyl)silane 93%
Trimethoxy(4-methoxyphenyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄Si
MDL Number: MFCD01861721
CAS Number: 35692-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR