2-Methyl-3-(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A475038-100G A475038-10G A475038-1G A475038-250MG A475038-25G A475038-500G A475038-5G
77279-386EA 347 USD
77279-386 77279-388 77279-390 77279-392 77279-394 77279-396 77279-398
2-Methyl-3-(trifluoromethyl)benzoic acid 95%
2-Methyl-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MDL Number: MFCD01631588
CAS Number: 62089-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR