Fmoc-NH-PEG5-CH2COOH 96%

Supplier: Ambeed
Warning

A474989-100MG A474989-1G A474989-250MG
77265-414EA 37 USD
77265-414 77265-416 77748-985
Fmoc-NH-PEG5-CH2COOH 96%
Fmoc-NH-PEG5-CH2COOH

Precaución: for Research Use Only

Formula: C₂₇H₃₅NO₉
MDL Number: MFCD26142982
CAS Number: 635287-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR