9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(1,3,2-dioxaborinane)
A474547-1G
A474547-250MG
A474547-25G
A474547-5G
77241-268EA
41
USD
77241-268
77241-270
77241-272
77746-972
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester 98%
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester
Precaución: for Research Use Only
Formula:
C₃₅H₅₂B₂O₄ |
MDL Number:
MFCD03701607 CAS Number: 317802-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...