Methyl 6-bromo-4-chloropicolinate 97%

Supplier: Ambeed
Warning

A474007-100MG A474007-250MG A474007-50MG
77241-262EA 173.04 USD
77241-262 77241-264 77241-266
Methyl 6-bromo-4-chloropicolinate 97%
Methyl 6-bromo-4-chloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.47 g/mol
MDL Number: MFCD14582022
CAS Number: 1206249-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR