4-Methyl-2H-1,4-benzoxazin-3(4H)-one 97%

Supplier: Ambeed
Warning

A473562-1G A473562-25G A473562-5G
77291-888EA 18 USD
77291-888 77291-890 77291-892
4-Methyl-2H-1,4-benzoxazin-3(4H)-one 97%
4-Methyl-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD00601365
CAS Number: 21744-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR