4-Anilino-1,1':4',1''-terphenyl 98%
Supplier: Ambeed
|
Warning
|
A473466-1G
A473466-25G
A473466-5G
77241-250EA
16
USD
77241-250
77241-252
77241-254
4-Anilino-1,1':4',1''-terphenyl 98%
4-Anilino-1,1':4',1''-terphenyl
Precaución: for Research Use Only
Formula:
C₂₄H₁₉N |
MDL Number:
MFCD28963769 CAS Number: 897671-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...