4-Amino-1-methylpyrimidin-2(1H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylcytosine

A473407-5G A473407-1G A473407-100MG
77241-248EA 1614 USD
77241-248 77746-970 77746-969
4-Amino-1-methylpyrimidin-2(1H)-one 97%
4-Amino-1-methylpyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MDL Number: MFCD00038058
CAS Number: 1122-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR