2-Bromo-p-xylene 98%

Supplier: Ambeed
Warning

A472948-1000G A472948-100G A472948-10G A472948-25G A472948-500G
77265-382EA 153 USD
77265-382 77265-384 77265-386 77265-388 77265-390
2-Bromo-p-xylene 98%
2-Bromo-p-xylene

Precaución: for Research Use Only

Formula: C₈H₉Br
MDL Number: MFCD00000074
CAS Number: 553-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR