Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: Cyclopentadienyl(p-cymene) ruthenium(II)hexafluorophosphate

A472916-100MG A472916-1G A472916-250MG
77253-058EA 53 USD
77253-058 77253-060 77253-062
Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate 98%
Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₅H₁₉F₆PRu
MW: 445.3500 g/mol
MDL Number: MFCD06798307
CAS Number: 147831-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR