1-Thien-2-ylbutan-1-one 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Thiophen-2-yl)butan-1-one

A472887-100G A472887-25G A472887-5G
77279-376EA 76.22 USD
77279-376 77279-378 77279-380
1-Thien-2-ylbutan-1-one 98%
1-Thien-2-ylbutan-1-one

Precaución: for Research Use Only

Formula: C₈H₁₀OS
MW: 154.23 g/mol
MDL Number: MFCD00022503
CAS Number: 5333-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR