2,6-Dimethyl-4-pyrone 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethyl-4H-pyran-4-one, 2,6-Dimethyl-γ-pyrone

A472870-100G A472870-1G A472870-25G A472870-500G A472870-5G
77253-048EA 92 USD
77253-048 77253-050 77253-052 77253-054 77253-056
2,6-Dimethyl-4-pyrone 95%
2,6-Dimethyl-4-pyrone

Precaución: for Research Use Only

Formula: C₇H₈O₂
MDL Number: MFCD00006579
CAS Number: 1004-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR