5,9-Dibromo-7,7-diphenyl-7H-benzo[c]fluorene 97%

Supplier: Ambeed
Warning

A472794-100MG A472794-1G A472794-250MG
77265-376EA 28.84 USD
77265-376 77265-378 77265-380
5,9-Dibromo-7,7-diphenyl-7H-benzo[c]fluorene 97%
5,9-Dibromo-7,7-diphenyl-7H-benzo[c]fluorene

Precaución: for Research Use Only

Formula: C₂₉H₁₈Br₂
MW: 526.26 g/mol
MDL Number: MFCD27979163
CAS Number: 854952-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR