4-Amino-2-methylquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminoquinaldine, 2-Methylquinolin-4-amine

A472719-1G A472719-25G A472719-5G
77291-864EA 31.93 USD
77291-864 77291-866 77291-868
4-Amino-2-methylquinoline 98%
4-Amino-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
MDL Number: MFCD00006759
CAS Number: 6628-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR