IDO-IN-2 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(Diisobutylamino)-2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-3-yl)-3-(p-tolyl)urea, PCC0208009

A472667-10MG A472667-1MG A472667-25MG A472667-50MG A472667-5MG
77265-366EA 53.56 USD
77265-366 77265-368 77265-370 77265-372 77265-374
IDO-IN-2 98+%
IDO-IN-2

Precaución: for Research Use Only

Formula: C₂₉H₃₅N₇O
MW: 497.65 g/mol
MDL Number: MFCD30728441
CAS Number: 1668565-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR