2-Chloroquinolin-7-amine 97%

Supplier: Ambeed
Warning

A472663-100MG A472663-1G A472663-250MG A472663-50MG
77265-358EA 185 USD
77265-358 77265-360 77265-362 77265-364
2-Chloroquinolin-7-amine 97%
2-Chloroquinolin-7-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
CAS Number: 1782402-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR