6-Bromo-3-methylpicolinic acid 97%

Supplier: Ambeed
Warning

A472589-1G A472589-5G A472589-100MG
77265-356EA 60 USD
77265-356 77572-486 77748-979
6-Bromo-3-methylpicolinic acid 97%
6-Bromo-3-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD16249865
CAS Number: 1211516-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR