3-Bromo-2-chloro-4-ethoxypyridine 97%

Supplier: Ambeed
Danger

A472117-1G A472117-25G A472117-5G
77265-344EA 40.17 USD
77265-344 77265-346 77265-348
3-Bromo-2-chloro-4-ethoxypyridine 97%
3-Bromo-2-chloro-4-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MW: 236.49 g/mol
MDL Number: MFCD00234276
CAS Number: 52311-47-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR