RGFP966 98%

Supplier: Ambeed
Warning

A471979-25MG A471979-50MG
77241-240EA 130 USD
77241-240 77241-242
RGFP966 98%
RGFP966

Precaución: for Research Use Only

Formula: C₂₁H₁₉FN₄O
MDL Number: MFCD26960954
CAS Number: 1357389-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR