GW5074 98%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-3-[(3,5-dibromo-4-hydroxyphenyl)methylene]-2-indolinone, 3-(3,5-dibromo-4-hydroxybenzylidene)-5-iodoindolin-2-one, GW 5074

A471753-10MG A471753-1MG A471753-50MG A471753-100MG A471753-1G A471753-250MG
77241-234EA 33.99 USD
77241-234 77241-236 77241-238 77617-048 77746-966 77746-967
GW5074 98%
GW5074

Precaución: for Research Use Only

Formula: C₁₅H₈Br₂INO₂
MW: 520.94 g/mol
MDL Number: MFCD09971042
CAS Number: 220904-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR