Glecaprevir 98%

Supplier: Ambeed
Warning

Synonyms: ABT-493, A 1282576, (33R,35S,91R,92R,5S,E)-5-(tert-Butyl)-N-((1R,2R)-2-(difluoromethyl)-1-(((1-methylcyclopropyl)sulphonyl)carbamoyl)cyclopropyl)-14,14-difluoro-4,7-dioxo-2,8,10-trioxa-6-aza-1(2,3)-quinoxalina-3(3,1)-pyrrolidina-9(1,2)-cyclopentanacyclotetradecaphan-12-ene-35-carboxamide

A471517-10MG A471517-1MG A471517-50MG A471517-5MG A471517-250MG A471517-100MG
77279-364EA 42.23 USD
77279-364 77279-366 77279-368 77279-370 77617-046 77748-976
Glecaprevir 98%
Glecaprevir

Precaución: for Research Use Only

Formula: C₃₈H₄₆F₄N₆O₉S
MW: 838.87 g/mol
MDL Number: MFCD30533436
CAS Number: 1365970-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR