3-(Butylcarbamoyl)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Butylaminocarbonyl)phenylboronic acid, 3-(n-Butylcarbamoyl)benzeneboronic acid, (3-(Butylcarbamoyl)phenyl)boronic acid
A470863-1G
A470863-250MG
A470863-25G
A470863-5G
77279-350EA
43
USD
77279-350
77279-352
77279-354
77279-356
3-(Butylcarbamoyl)phenylboronic acid 98%
3-(Butylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₆BNO₃ |
MDL Number:
MFCD04115691 CAS Number: 397843-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...