(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%
Supplier: Ambeed
|
Warning
|
A469348-100MG
A469348-1G
A469348-250MG
77253-034EA
74
USD
77253-034
77253-036
77253-038
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%
(R)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate
Precaución: for Research Use Only
Formula:
C₃₆H₂₅F₁₂O₄P |
MDL Number:
MFCD30489114 CAS Number: 1011465-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...