3-Iodo-5-azaindole 95%

Supplier: Ambeed
Warning

A468942-100MG A468942-1G A468942-250MG A468942-5G
77279-332EA 12.36 USD
77279-332 77279-334 77279-336 77279-338
3-Iodo-5-azaindole 95%
3-Iodo-5-azaindole

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MW: 244.03 g/mol
MDL Number: MFCD08234839
CAS Number: 877060-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR